Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MTHFD2L Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518916
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
The specific function of this protein remains unknown.
Specifications
MTHFD2L | |
Polyclonal | |
Unconjugated | |
MTHFD2L | |
1110019K23Rik; C630010D07Rik; FLJ13105; LOW QUALITY PROTEIN: probable bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase 2; Methenyltetrahydrofolate cyclohydrolase; methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2 like; methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2-like; methylenetetrahydrofolate dehydrogenase 2-like; MGC45532; MGC72244; MTHFD2L; MTHFD2-like; NAD-dependent methylenetetrahydrofolate dehydrogenase; NADP-dependent methylenetetrahydrofolate dehydrogenase 2-like protein; probable bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase 2; probable bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase 2; methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2-like; probable bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase 2-like; putative bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase 2-like protein; RGD1310879; tetrahydrofolate dehydrogenase/cyclohydrolase; Unknown (protein for MGC:63479); wu:fi14c12; zgc:63479 | |
Goat | |
Ammonium Sulfate Precipitation | |
RUO | |
441024 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9H903 | |
MTHFD2L | |
Synthetic peptide sequence (KHIQKEIQRGVES) corresponding to the N-terminus amino acids of MTHFD2L (aa 61-74). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction