Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MTHFR Antibody (5D3), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP237607
Description
MTHFR Monoclonal specifically detects MTHFR in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MTHFR | |
Monoclonal | |
Unconjugated | |
Ascites with 0.03% Sodium Azide | |
MTHFR | |
Purified recombinant fragment of human MTHFR expressed in E. Coli. | |
Unpurified | |
RUO | |
4524 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
5D3 | |
Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin | |
EC 1.5.1.20, methylenetetrahydrofolate reductase, methylenetetrahydrofolate reductase (NAD(P)H)5,10-methylenetetrahydrofolate reductase (NADPH) | |
Mouse | |
75 kDa | |
0.1 mL | |
Primary | |
Human, Rat | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction