Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTHFR Monoclonal Antibody (5D3)
GREENER_CHOICE

Catalog No. PIMA515844
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515844 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515844 Supplier Invitrogen™ Supplier No. MA515844
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15844 targets MTHFR in indirect ELISA, IHC and WB applications and shows reactivity with Human and Rat samples. The MA5-15844 immunogen is purified recombinant fragment of human MTHFR expressed in E. Coli. . MA5-15844 detects MTHFR which has a predicted molecular weight of approximately 75kDa.

The protein encoded by this gene catalyzes the conversion of 5,10-methylenetetrahydrofolate to 5-methyltetrahydrofolate, a co-substrate for homocysteine remethylation to methionine. Genetic variation in this gene influences susceptibility to occlusive vascular disease, neural tube defects, colon cancer and acute leukemia, and mutations in this gene are associated with methylenetetrahydrofolate reductase deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTHFR
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 5D3
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene MTHFR
Gene Accession No. P42898
Gene Alias 5,10-methylenetetrahydrofolate reductase; 5,10-methylenetetrahydrofolate reductase (NADPH); AI323986; Methylenetetrahydrofolate reductase; methylenetetrahydrofolate reductase (NAD(P)H); MTHFR; RP11-56N19.4
Gene Symbols MTHFR
Host Species Mouse
Immunogen Purified recombinant fragment of human MTHFR expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362657, 4524
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.