Learn More
MTHFR Mouse anti-Human, Clone: OTI2H3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF812635

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene catalyzes the conversion of 5,10-methylenetetrahydrofolate to 5-methyltetrahydrofolate, a co-substrate for homocysteine remethylation to methionine. Genetic variation in this gene influences susceptibility to occlusive vascular disease, neural tube defects, colon cancer and acute leukemia, and mutations in this gene are associated with methylenetetrahydrofolate reductase deficiency.Specifications
MTHFR | |
Monoclonal | |
Unconjugated | |
MTHFR | |
methylenetetrahydrofolate reductase | |
Mouse | |
Affinity Chromatography | |
RUO | |
4524 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2H3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P42898 | |
MTHFR | |
Human recombit protein fragment corresponding to amino acids 1-230 of human MTHFR produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.