Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTHFR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142473
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142473 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142473 Supplier Thermo Scientific Supplier No. PA5142473
Add to Cart
Add to Cart
This item is not returnable. View return policy

MTHFR Polyclonal Antibody

MTHFR Polyclonal Antibody for IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen MTHFR
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene MTHFR
Gene Accession No. P42898
Gene Alias 5,10-methylenetetrahydrofolate reductase; 5,10-methylenetetrahydrofolate reductase (NADPH); AI323986; Methylenetetrahydrofolate reductase; methylenetetrahydrofolate reductase (NAD(P)H); MTHFR; RP11-56N19.4
Gene Symbols MTHFR
Host Species Goat
Immunogen Peptide with sequence C-NRPTQNARETEAP (aa 644-656).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4524
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.