Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTHFSD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559520
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559520 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559520 Supplier Invitrogen™ Supplier No. PA559520
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VSKQDIREQI WGYMESQNLA DFPRPVHHRI PNFKGSYLAC QNIKDLDVFA RTQEVKVDPD KPLEGVRLLV LQSKKTLLVP TPRLRTGLFN KITP Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 63%.

MTHFSD (Methenyltetrahydrofolate Synthetase Domain Containing) is a Protein Coding gene. Diseases associated with MTHFSD include Pancreas, Annular and Anus, Imperforate. Gene Ontology (GO) annotations related to this gene include nucleic acid binding and nucleotide binding. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTHFSD
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MTHFSD
Gene Accession No. Q2M296
Gene Alias AW049977; BC052066; C920004D05; fc43c12; LOW QUALITY PROTEIN: methenyltetrahydrofolate synthase domain-containing protein; methenyltetrahydrofolate synthase domain-containing protein; methenyltetrahydrofolate synthase domain-containing protein; LOW QUALITY PROTEIN: methenyltetrahydrofolate synthase domain-containing protein; methenyltetrahydrofolate synthetase domain containing; methenyltetrahydrofolate synthetase domain-containing protein; MGC137477 protein; Mthfsd; si:dkey-199i24.1; Unknown (protein for MGC:153374); wu:fc43c12; zgc:153374
Gene Symbols MTHFSD
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MTHFSD. Recombinant protein control fragment (Product #RP-98625).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64779
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.