Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTIF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548811
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548811 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548811 Supplier Invitrogen™ Supplier No. PA548811
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MTIF2 Polyclonal Antibody for Western Blot

One of the essential components for the initiation of protein synthesis. Protects formylmethionyl-tRNA from spontaneous hydrolysis and promotes its binding to the 30S ribosomal subunits. Also involved in the hydrolysis of GTP during the formation of the 70S ribosomal complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTIF2
Applications Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene MTIF2
Gene Accession No. P46199
Gene Alias IF2(mt); IF-2(Mt); IF-2mt; mitochondrial translational initiation factor 2; MTIF2; Translation initiation factor IF-2, mitochondrial
Gene Symbols MTIF2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 138-167 amino acids from the N-terminal region of human MTIF2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4528
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.