Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTIF3 Monoclonal Antibody (OTI1E8)
GREENER_CHOICE

Catalog No. PIMA526438
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526438 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526438 Supplier Invitrogen™ Supplier No. MA526438
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MTIF3 Monoclonal Antibody for Western Blot, IHC (P)

This gene encodes a translation initiation factor that is involved in mitochondrial protein synthesis. Polymorphism in this gene is associated with the onset of Parkinson's disease. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTIF3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1E8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MTIF3
Gene Accession No. Q9CZD5, Q9H2K0
Gene Alias 2810012L14Rik; AI414549; DC38; IF3(mt); IF-3(Mt); IF3mt; IF-3Mt; mitochondrial translational initiation factor 3; Mtif3; Translation initiation factor IF-3, mitochondrial
Gene Symbols MTIF3
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 56-278 of MTIF3 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219402, 477328, 76366
Target Species Human, Mouse, Canine, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.