Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTIF3 Monoclonal Antibody (OTI4B6)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA526442

Catalog No. PIMA526442


Only null left
Add to Cart

Description

Description

MTIF3 Monoclonal Antibody for Western Blot, IHC (P)

This gene encodes a translation initiation factor that is involved in mitochondrial protein synthesis. Polymorphism in this gene is associated with the onset of Parkinson's disease. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 5.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MTIF3
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9H2K0
MTIF3
Human recombinant protein fragment corresponding to amino acids 56-278 of MTIF3 produced in E.coli.
100 μL
Primary
Human, Canine, Monkey
Antibody
IgG2a
Immunohistochemistry (Paraffin), Western Blot
OTI4B6
Unconjugated
MTIF3
2810012L14Rik; AI414549; DC38; IF3(mt); IF-3(Mt); IF3mt; IF-3Mt; mitochondrial translational initiation factor 3; Mtif3; Translation initiation factor IF-3, mitochondrial
Mouse
Affinity Chromatography
RUO
219402, 477328
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.