Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTIF3 Mouse anti-Canine, Human, Mouse, Clone: OTI1E8, liquid, TrueMAB™
SDP

Catalog No. 501678308
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-8308 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-8308 Supplier OriGene Supplier No. TA800421
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This gene encodes a translation initiation factor that is involved in mitochondrial protein synthesis. Polymorphism in this gene is associated with the onset of Parkinson's disease. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 5.

Specifications

Antigen MTIF3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1E8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MTIF3
Gene Accession No. Q9CZD5, Q9H2K0
Gene Alias IF3mt
Gene Symbols MTIF3
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 56-278 of human MTIF3 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219402, 477328, 76366
Target Species Canine, Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.