Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTIF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558930
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558930 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558930 Supplier Invitrogen™ Supplier No. PA558930
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KHLVQITIKK GKNVDVSENE MEEIFHQILQ TMPGIATFSS RPQAVQGGKA LMCVLRALSK NEEKAYKETQ ETQERDTLNK DHGNDKESNV LHQ Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 61%.

This gene encodes a translation initiation factor that is involved in mitochondrial protein synthesis. Polymorphism in this gene is associated with the onset of Parkinson's disease. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTIF3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MTIF3
Gene Accession No. Q9H2K0
Gene Alias 2810012L14Rik; AI414549; DC38; IF3(mt); IF-3(Mt); IF3mt; IF-3Mt; mitochondrial translational initiation factor 3; Mtif3; Translation initiation factor IF-3, mitochondrial
Gene Symbols MTIF3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MTIF3. Recombinant protein control fragment (Product #RP-97936).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219402
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.