Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589279
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589279 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589279 Supplier Invitrogen™ Supplier No. PA589279
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SARERQKVTE RTVSLWSLIN SNKEKFKNPF YTKEINRVLY PVASMRHLEL WVNYYIRWNP RIKQQQPNPV EQRYMELLAL RDEYIKRLEE LQLANSAKLS DPPTSPSSPS QMMPHVQTHF; Positive Samples: Mouse thymus; Cellular Location: Cell membrane, Cell projection, Cytoplasm, Late endosome, Peripheral membrane protein, filopodium, ruffle.

Human MTM1, a 603 amino-acid protein, is mutated in myotubular myopathy. The largely related protein hMTMR2 is found mutated in a recessive form of Charcot-Marie-tooth neuropathy. Myotubularin is primarily a lipid phosphatase that acts on hosphatidylinositol 3-monophosphate and is involved in the regulation of the phosphatidylinositol 3-kinase (PI 3-kinase) pathway and membrane trafficking. Wildtype myotubularin can directly dephosphorylate PI 3-P and PI 4-P in vitro. Thus, it decreases PI 3-P levels by down-regulating PI 3-K activity and by facilitating the degradation of PI 3-P.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTM1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.14 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MTM1
Gene Accession No. Q6AXQ4, Q9Z2C5
Gene Alias AF073996; CG2; CNM; fb19c01; hypothetical protein LOC560881; im:6903724; mKIAA4176; Mtm; MTM1; MTMX; myotubularin; myotubularin 1; Myotubularin-like protein; phosphatidylinositol-3,5-bisphosphate 3-phosphatase; Phosphatidylinositol-3-phosphate phosphatase; wu:fb19c01; X-linked myotubular myopathy gene 1; XLMTM; zgc:123266
Gene Symbols MTM1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 484-603 of human MTM1 (NP_0002431).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17772, 288762
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.