Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517972
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517972 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517972 Supplier Thermo Scientific Supplier No. PA517972
Add to Cart
Add to Cart

MTM1 Polyclonal Antibody

MTM1 Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen MTM1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene MTM1
Gene Accession No. Q13496, Q9Z2C5
Gene Alias AF073996; CG2; CNM; fb19c01; hypothetical protein LOC560881; im:6903724; mKIAA4176; Mtm; MTM1; MTMX; myotubularin; myotubularin 1; Myotubularin-like protein; phosphatidylinositol-3,5-bisphosphate 3-phosphatase; Phosphatidylinositol-3-phosphate phosphatase; wu:fb19c01; X-linked myotubular myopathy gene 1; XLMTM; zgc:123266
Gene Symbols MTM1
Host Species Goat
Immunogen Synthetic peptide sequence (SSPSQMMPHVQTHF) corresponding to the C-terminus amino acids of MTM1 (aa 590-603).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17772, 4534
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.