Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MTM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552725
Description
Immunogen sequence: LDHLYSCRFG TFLFNCESAR ERQKVTERTV SLWSLINSNK EKFKNPFYTK EINRVLYPVA SMRHLELWVN YYIRWNPRIK QQQPNPVEQR YMELLALRDE YIKRLEELQL ANSAKLSDPP TSPSSPSQMM P Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 55%.
Human MTM1, a 603 amino-acid protein, is mutated in myotubular myopathy. The largely related protein hMTMR2 is found mutated in a recessive form of Charcot-Marie-tooth neuropathy. Myotubularin is primarily a lipid phosphatase that acts on hosphatidylinositol 3-monophosphate and is involved in the regulation of the phosphatidylinositol 3-kinase (PI 3-kinase) pathway and membrane trafficking. Wildtype myotubularin can directly dephosphorylate PI 3-P and PI 4-P in vitro. Thus, it decreases PI 3-P levels by down-regulating PI 3-K activity and by facilitating the degradation of PI 3-P.
Specifications
MTM1 | |
Polyclonal | |
Unconjugated | |
MTM1 | |
AF073996; CG2; CNM; fb19c01; hypothetical protein LOC560881; im:6903724; mKIAA4176; Mtm; MTM1; MTMX; myotubularin; myotubularin 1; Myotubularin-like protein; phosphatidylinositol-3,5-bisphosphate 3-phosphatase; Phosphatidylinositol-3-phosphate phosphatase; wu:fb19c01; X-linked myotubular myopathy gene 1; XLMTM; zgc:123266 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4534 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13496 | |
MTM1 | |
Recombinant protein corresponding to Human MTM1. Recombinant protein control fragment (Product #RP-92237). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction