Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTMR1 Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB269777 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB269777 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB269777 Supplier Novus Biologicals Supplier No. NBP244289AF594
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MTMR1 Polyclonal antibody specifically detects MTMR1 in Human samples. It is validated for Western Blot

Specifications

Antigen MTMR1
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias EC 3.1.3, EC 3.1.3.-, EC 3.1.3.48, myotubularin related protein 1, myotubularin-related protein 1
Host Species Rabbit
Immunogen The epitope recognized by this antibody maps to a region between residue 615 to 665 of human Myotubularin-related protein 1 using the numbering given in entry Q13613.4 (GeneID 8776).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8776
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.