Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTMR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145121
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145121 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. FEPA5145121 missing translation for 'mfr' Invitrogen™ missing translation for 'supplierNo' PA5145121
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EGLQREVATR AVSSSSERGS SPSHSATSVH T Antigen sequence identity: Mouse 81%; Rat 71%.

MTMR1 is a member of the myotubularin related family of proteins. Members of this family contain the consensus sequence for the active site of protein tyrosine phosphatases.This gene encodes a member of the myotubularin related family of proteins. Members of this family contain the consensus sequence for the active site of protein tyrosine phosphatases. Alternatively spliced variants have been described but their biological validity has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTMR1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MTMR1
Gene Accession No. Q13613
Gene Alias AW049210; LOW QUALITY PROTEIN: myotubularin-related protein 1; Mtmr1; myotubularin related protein 1; myotubularin-related protein 1; Phosphatidylinositol-3,5-bisphosphate 3-phosphatase; phosphatidylinositol-3-phosphate phosphatase
Gene Symbols MTMR1
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 633-663. Recombinant protein control fragment (Product #RP-110137).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8776
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.