Learn More
MTMR1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304917AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
MTMR1 is a member of the myotubularin related family of proteins. Members of this family contain the consensus sequence for the active site of protein tyrosine phosphatases.This gene encodes a member of the myotubularin related family of proteins. Members of this family contain the consensus sequence for the active site of protein tyrosine phosphatases. Alternatively spliced variants have been described but their biological validity has not been determined.Specifications
MTMR1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q13613 | |
MTMR1 | |
Between 615 and 665 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
MTMR1 | |
Myotubularin related protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8776 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.