Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTMR12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562384
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562384 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562384 Supplier Invitrogen™ Supplier No. PA562384
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RPEEIHTNEK EVTEKEVTLH LLPGEQLLCE ASTVLKYVQE DSCQHGVYGR LVCTDFKIAF LGDDESALDN DETQFKNKVI GENDITLHCV DQIYGVFDE Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 85%.

Phosphatidylinositide 3-kinase-derived membrane-anchored phosphatidylinositides, such as phosphatidylinositol 3-phosphate (PtdIns(3)P), regulate diverse cellular processes. The protein encoded by this gene functions as an adaptor subunit in a complex with an active PtdIns(3)P 3-phosphatase. Alternatively spliced transcript variants have been identified, but their biological validity has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTMR12
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MTMR12
Gene Accession No. Q9C0I1
Gene Alias 3Pap; 3-PAP; 3-phosphatase adapter protein; 3-phosphatase adapter subunit; 4932703C11; C730015A02Rik; im:7160376; Inactive phosphatidylinositol 3-phosphatase 12; Kiaa1682; mKIAA1682; Mtmr12; myotubularin related protein 12; myotubularin-related protein 12; Phosphatidylinositol 3 phosphate 3-phosphatase adapter subunit; phosphatidylinositol 3-phosphatase-associated protein; phosphatidylinositol-3 phosphate 3-phosphatase adapter subunit; phosphatidylinositol-3 phosphate 3-phosphatase adaptor subunit; phosphatidylinositol-3-phosphatase associated protein; phosphatidylinositol-3-phosphatase-associated protein; phosphatidylinositol-3-phosphate associated protein; Pip3ap; si:dkey-207e22.3
Gene Symbols MTMR12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MTMR12. Recombinant protein control fragment (Product #RP-101360).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54545
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.