Learn More
Invitrogen™ MTMR14 Monoclonal Antibody (OTI6B6)

Description
MTMR14 Monoclonal Antibody for Western Blot, ICC/IF, Flow
Specifications
Specifications
| Antigen | MTMR14 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI6B6 |
| Concentration | 0.44 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | MTMR14 |
| Gene Accession No. | Q8NCE2 |
| Gene Alias | 1110061O04Rik; AW553738; C3orf29; C76151; egg-derived tyrosine phosphatase homolog; fc14d11; HCV NS5A-transactivated protein 4 splice variant A-binding protein 1; hJumpy; jumpy; mJumpy; Mtmr14; myotubularin related protein 14; myotubularin-related protein 14; NS5ATP4ABP1; RGD1304842; wu:fc14d11 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.