Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MTMR14 Monoclonal Antibody (OTI6B6), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA501774S


Description
MTMR14 Monoclonal antibody specifically detects MTMR14 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
MTMR14 | |
Monoclonal | |
0.44 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P17066 | |
Mouse | |
Affinity chromatography | |
RUO | |
3310 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Western Blot | |
OTI6B6 | |
Unconjugated | |
HSPA6 | |
1110061O04Rik; AW553738; C3orf29; C76151; egg-derived tyrosine phosphatase homolog; fc14d11; HCV NS5A-transactivated protein 4 splice variant A-binding protein 1; hJumpy; jumpy; mJumpy; Mtmr14; myotubularin related protein 14; myotubularin-related protein 14; NS5ATP4ABP1; RGD1304842; wu:fc14d11 | |
Full length human recombinant protein of human HSPA6 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction