Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTMR14 Monoclonal Antibody (OTI6F8)
GREENER_CHOICE

Catalog No. PIMA525219
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525219 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525219 Supplier Invitrogen™ Supplier No. MA525219
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MTMR14 Monoclonal Antibody for Western Blot, ICC/IF, Flow

This gene encodes a myotubularin-related protein. The encoded protein is a phosphoinositide phosphatase that specifically dephosphorylates phosphatidylinositol 3,5-biphosphate and phosphatidylinositol 3-phosphate. Mutations in this gene are correlated with autosomal dominant centronuclear myopathy. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 18.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTMR14
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI6F8
Concentration 0.44 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MTMR14
Gene Accession No. Q8NCE2
Gene Alias 1110061O04Rik; AW553738; C3orf29; C76151; egg-derived tyrosine phosphatase homolog; fc14d11; HCV NS5A-transactivated protein 4 splice variant A-binding protein 1; hJumpy; jumpy; mJumpy; Mtmr14; myotubularin related protein 14; myotubularin-related protein 14; NS5ATP4ABP1; RGD1304842; wu:fc14d11
Gene Symbols MTMR14
Host Species Mouse
Immunogen Full length human recombinant protein of MTMR14 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64419
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.