Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTMR14 Monoclonal Antibody (OTI9G9), TrueMAB™, OriGene
SDP

Mouse Monoclonal Antibody

$231.00 - $516.29

Specifications

Antigen MTMR14
Clone OTI9G9
Concentration 0.96 mg/mL
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-254-2077
SDP
Encompass
Encompass_Preferred
missing translation for 'viewDocuments'
OriGene
TA501712S
30 μL
Each for $231.00
Only null left
Add to Cart
 
50-167-2831
SDP
Encompass_Preferred
missing translation for 'viewDocuments'
OriGene
TA501712
100 μL
Each for $516.29
Only null left
Add to Cart
 
Description

Description

MTMR14 Monoclonal antibody specifically detects MTMR14 in Canine, Human, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot
Specifications

Specifications

MTMR14
0.96 mg/mL
Unconjugated
Mouse
IgG2b
-20° C, Avoid Freeze/Thaw Cycles
OTI9G9
Monoclonal
Liquid
RUO
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.