Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTMR15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112863
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112863 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112863 Supplier Invitrogen™ Supplier No. PA5112863
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MTMR15 Polyclonal Antibody for Western Blot, IHC (P), ELISA

MTMRF encodes a member of the myotubularin-related class 1 cysteine-based protein tyrosine phosphatases. The encoded protein may be catalytically inactive.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTMR15
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene FAN1
Gene Accession No. Q9Y2M0
Gene Alias coiled-coil domain-containing protein MTMR15; FAN1; FANCD2/FANCI-associated nuclease 1; fanconi anemia associated nuclease 1; fanconi-associated nuclease 1; hFAN1; KIAA1018; KMIN; MTMR15; MTMRF; Myotubularin-related protein 15
Gene Symbols FAN1
Host Species Rabbit
Immunogen Recombinant Human Fanconi-associated nuclease 1 protein (167-373AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22909
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.