Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MTMR2 Antibody (OTI1F10), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272810B
Description
MTMR2 Monoclonal specifically detects MTMR2 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot.Specifications
MTMR2 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
MTMR2 | |
Full length recombinant protein of human MTMR2 (NP_057240) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
IgG1 |
Western Blot | |
OTI1F10 | |
Western Blot | |
CMT4B, CMT4B1, EC 3.1.3, EC 3.1.3.2, EC 3.1.3.48, KIAA1073EC 3.1.3.-, myotubularin related protein 2, myotubularin-related protein 2 | |
Mouse | |
Affinity Purified | |
Cancer, Protein Phosphatase | |
8898.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction