Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MTMR2 Antibody (OTI2D2) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272809
Description
MTMR2 Monoclonal specifically detects MTMR2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
MTMR2 | |
Monoclonal | |
LYOPH | |
Western Blot 1:2000, Flow Cytometry 1:100, CyTOF-ready | |
CMT4B, CMT4B1, EC 3.1.3, EC 3.1.3.2, EC 3.1.3.48, KIAA1073EC 3.1.3.-, myotubularin related protein 2, myotubularin-related protein 2 | |
Mouse | |
73.2 kDa | |
100 μg | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Flow Cytometry, CyTOF | |
OTI2D2 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
MTMR2 | |
Full length human recombinant protein of human MTMR2 (NP_057240) produced in HEK293T cell. | |
Affinity Purified | |
Cancer, Protein Phosphatase | |
8898 | |
Human, Mouse, Rat | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction