Learn More
MTMR2 Mouse anti-Human, Clone: OTI5G5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502204

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene is a member of the myotubularin family. The encoded protein possesses phosphatase activity towards phosphatidylinositol-3-phosphate and phosphatidylinositol-3,5-bisphosphate. Mutations in this gene are a cause of Charcot-Marie-Tooth disease type 4B, an autosomal recessive demyelinating neuropathy. Multiple alternatively spliced transcript variants have been found for this gene.Specifications
MTMR2 | |
Monoclonal | |
Unconjugated | |
MTMR2 | |
CMT4B, CMT4B1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
8898 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
OTI5G5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q13614 | |
MTMR2 | |
Full length human recombit protein of human MTMR2 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.