Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTMR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597174
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597174 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597174 Supplier Invitrogen™ Supplier No. PA597174
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DKVESGKTSV VVHCSDGWDR TAQLTSLAML MLDGYYRTIR GFEVLVEKEW LSFGHRFQLR VGHGDKNHAD ADRSPVFLQF IDCVWQMTRQ FPTAFEFNEY FLITILDHLY SCLFGTFLCN SEQQRGKENL PKRTVSLWSY INSQLEDFTN PLYGSYSNHV LYPVASMRHL ELWVGYYIRW NPRMKPQEPI HNRYKELLAK RAELQKKVEE LQREISNRST SSSERASSPA QCVTPVQTVV; Positive Samples: HeLa, Jurkat, U-87MG, Mouse brain, Mouse kidney, Mouse liver, Rat liver; Cellular Location: Cytoplasm, Early endosome membrane, Peripheral membrane protein.

This gene is a member of the myotubularin family. The encoded protein possesses phosphatase activity towards phosphatidylinositol-3-phosphate and phosphatidylinositol-3,5-bisphosphate. Mutations in this gene are a cause of Charcot-Marie-Tooth disease type 4B, an autosomal recessive demyelinating neuropathy. Multiple alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTMR2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.09 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MTMR2
Gene Accession No. Q13614, Q9Z2D1
Gene Alias 6030445P13Rik; CMT4B; CMT4B1; KIAA1073; Mtmr2; myotubularin related protein 2; myotubularin-related protein 2; Phosphatidylinositol-3,5-bisphosphate 3-phosphatase; phosphatidylinositol-3-phosphatase; phosphatidylinositol-3-phosphate phosphatase; phosphoinositide-3-phosphatase
Gene Symbols MTMR2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 404-643 of human MTMR2 (NP_0572403).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 315422, 77116, 8898
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.