Learn More
Invitrogen™ MTMR3 Monoclonal Antibody (1E11)

Description
Peptide Sequence: CPSPTTPVDD SCAPYPAPGT SPDDPPLSRL PKTRSYDNLT TACDNTVPLA SRRCSDPSLN EKWQEHRRSL ELSSLAGPGE DPLSADSLGK PTRVPG.
Specifications
Specifications
| Antigen | MTMR3 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 1E11 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.2 |
| Gene | MTMR3 |
| Gene Accession No. | Q13615 |
| Gene Alias | 1700092A20Rik; AI255150; AW557713; FYVE (Fab1 YGLO23 Vsp27 EEA1 domain) dual-specificity protein phosphatase; FYVE domain-containing dual specificity protein phosphatase 1; FYVE-DSP1; KIAA0371; mKIAA0371; MTMR3; myotubularin related protein 3; myotubularin-related protein 3; Phosphatidylinositol-3,5-bisphosphate 3-phosphatase; phosphatidylinositol-3-phosphate phosphatase; ZFYVE10; zinc finger FYVE domain-containing protein 10; zinc finger, FYVE domain containing 10 |
| Gene Symbols | MTMR3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.