Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTMR3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556961
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556961 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556961 Supplier Invitrogen™ Supplier No. PA556961
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEPSPSETSL VERPQVGSVV HRTSLGSTLS LTRSPCALPL AECKEGLVCN GAPETENRAS EQPPGLSTLQ MYPTPNGHCA NGEAGRSKDS LSR Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 66%.

This gene encodes a member of the myotubularin dual specificity protein phosphatase gene family. The encoded protein is structurally similar to myotubularin but in addition contains a FYVE domain and an N-terminal PH-GRAM domain. The protein can self-associate and also form heteromers with another myotubularin related protein. The protein binds to phosphoinositide lipids through the PH-GRAM domain, and can hydrolyze phosphatidylinositol(3)-phosphate and phosphatidylinositol(3,5)-bisphosphate in vitro. The encoded protein has been observed to have a perinuclear, possibly membrane-bound, distribution in cells, but it has also been found free in the cytoplasm. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTMR3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MTMR3
Gene Accession No. Q13615
Gene Alias 1700092A20Rik; AI255150; AW557713; FYVE (Fab1 YGLO23 Vsp27 EEA1 domain) dual-specificity protein phosphatase; FYVE domain-containing dual specificity protein phosphatase 1; FYVE-DSP1; KIAA0371; mKIAA0371; MTMR3; myotubularin related protein 3; myotubularin-related protein 3; Phosphatidylinositol-3,5-bisphosphate 3-phosphatase; phosphatidylinositol-3-phosphate phosphatase; ZFYVE10; zinc finger FYVE domain-containing protein 10; zinc finger, FYVE domain containing 10
Gene Symbols MTMR3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MTMR3. Recombinant protein control fragment (Product #RP-95357).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8897
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.