Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTMR4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116250
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116250 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116250 Supplier Invitrogen™ Supplier No. PA5116250
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MTMR4 Polyclonal Antibody for Western Blot

MTMR, a member of the myotubularin family, encodes an intracellular protein of 1, 195 amino acids, and shares approximately 47% sequence identity with MTMR3, spiking to over 70% identity in the catalytic domain and the double zinc finger (FYVE) motif. PCR analysis detects expression of MTMR4 in all human tissues examined except lung, small intestine, stomach, salivary gland, adrenal gland, and uterus. Subcellular localization of MTMR4 in transfected HeLa cells showed a perinuclear distribution, similar to that seen with MTMR3. MTMR4 dephosphorylates para-nitrophenylphosphate and phosphatidylinositol 3-phosphate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTMR4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene MTMR4
Gene Accession No. Q9NYA4
Gene Alias AA596759; DKFZp781E194; ESTM44; FYVE domain-containing dual specificity protein phosphatase 2; FYVE zinc finger phosphatase; FYVE-DSP2; KIAA0647; MGC163449; MGC163451; mKIAA0647; Mtmr4; MTMR7; myotubularin related protein 4; myotubularin-related protein 4; ZFYVE11; Zinc finger FYVE domain-containing protein 11; zinc finger, FYVE domain containing 11
Gene Symbols MTMR4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 695-723 amino acids from the Central region of human MTMR4.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9110
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.