Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MTMR4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | MTMR4 |
|---|---|
| Concentration | 0.15 mg/mL |
| Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-8779
![]()
|
Proteintech Group Inc
134881AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
MTMR, a member of the myotubularin family, encodes an intracellular protein of 1, 195 amino acids, and shares approximately 47% sequence identity with MTMR3, spiking to over 70% identity in the catalytic domain and the double zinc finger (FYVE) motif. PCR analysis detects expression of MTMR4 in all human tissues examined except lung, small intestine, stomach, salivary gland, adrenal gland, and uterus. Subcellular localization of MTMR4 in transfected HeLa cells showed a perinuclear distribution, similar to that seen with MTMR3. MTMR4 dephosphorylates para-nitrophenylphosphate and phosphatidylinositol 3-phosphate.Specifications
| MTMR4 | |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q91XS1, Q9NYA4 | |
| 170749, 287607, 9110 | |
| MTMR4 Fusion Protein Ag4310 | |
| Primary | |
| -20°C | |
| MTMR4 |
| 0.15 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| FYVE DSP2, KIAA0647, MTMR4, myotubularin related protein 4, ZFYVE11 | |
| Mtmr4 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title