Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTMR6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590399
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590399 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590399 Supplier Invitrogen™ Supplier No. PA590399
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RCGQLDGDPK EVSPVFTQFL ECVWHLTEQF PQAFEFSEAF LLQIHEHIHS CQFGNFLGNC QKEREELKLK EKTYSLWPFL LEDQKKYLNP LYSSESHRFT VLEPNTVSFN FKFWRNMYHQ FDRTLHPRQS VFNIIMNMNE QNKQLEKDIK DLESKIKQRK NKQTDGILTK ELLHSVHPES PNLKTSLCFK EQTLLPVNDA LRTIEGSSPA DNRYSEYAEE FSKSEPAVVS LEYGVARMTC; Positive Samples: HepG2, HeLa, 293T, Mouse heart, Mouse brain, Mouse lung, Mouse spleen, Mouse testis, Rat heart; Cellular Location: Nucleus envelope.

Phosphatase that acts on lipids with a phosphoinositol headgroup (PubMed:19038970, PubMed:22647598). Dephosphorylates phosphatidylinositol 3-phosphate (PtdIns(3)P) and phosphatidylinositol 3,5-bisphosphate (PubMed:19038970, PubMed:22647598) (Probable). Binds with high affinity to phosphatidylinositol 3,5-bisphosphate (PtdIns(3,5)P2) but also to phosphatidylinositol 3-phosphate (PtdIns(3)P), phosphatidylinositol 4-phosphate (PtdIns(4)P), and phosphatidylinositol 5-phosphate (PtdIns(5)P), phosphatidic acid and phosphatidylserine (PubMed:19038970). Negatively regulates ER-Golgi protein transport. Probably in association with MTMR9, plays a role in the late stages of macropinocytosis by dephosphorylating phosphatidylinositol 3-phosphate in membrane ruffles (PubMed:24591580). Acts as a negative regulator of KCNN4/KCa3.1 channel activity in CD4(+) T-cells possibly by decreasing intracellular levels of phosphatidylinositol 3-phosphate (PubMed:15831468). Negatively regulates proliferation of reactivated CD4(+) T-cells (PubMed:16847315). In complex with MTMR9, negatively regulates DNA damage-induced apoptosis (PubMed:19038970, PubMed:22647598). The formation of the MTMR6-MTMR9 complex stabilizes both MTMR6 and MTMR9 protein levels (PubMed:19038970). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTMR6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.02 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MTMR6
Gene Accession No. Q8VE11, Q9Y217
Gene Alias 4022440C11Rik; AI428804; AU041072; MTMR6; myotubularin related protein 6; Myotubularin-like protein 6-like protein; myotubularin-related protein 6; Phosphatidylinositol-3,5-bisphosphate 3-phosphatase; Phosphatidylinositol-3-phosphate phosphatase
Gene Symbols MTMR6
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219135, 305935, 9107
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.