Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTMR7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114266
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114266 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114266 Supplier Invitrogen™ Supplier No. PA5114266
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NLKSSDPDLS ANSDQESGVE DLSCRSPSGG EHAPSEDSGK DRDSDEAVFL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 86%; Dog: 93%; Guinea Pig: 92%; Horse: 86%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

This gene encodes a member of the myotubularin family of tyrosine/dual-specificity phosphatases. The encoded protein is characterized by four distinct domains that are conserved among all members of the myotubularin family: the glucosyltransferase, Rab-like GTPase activator and myotubularins domain, the Rac-induced recruitment domain, the protein tyrosine phosphatases and dual-specificity phosphatases domain and the suppressor of variegation 3-9, enhancer-of-zeste, and trithorax interaction domain. This protein dephosphorylates the target substrates phosphatidylinositol 3-phosphate and inositol 1, 3-bisphosphate. A pseudogene of this gene is found on chromosome 5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTMR7
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Mtmr7
Gene Accession No. Q9Y216, Q9Z2C9
Gene Alias DKFZp781E194; Inositol 1,3-bisphosphate phosphatase; MGC163449; MGC163451; MTMR7; myotubularin related protein 7; myotubularin-related protein 7; Phosphatidylinositol-3-phosphate phosphatase
Gene Symbols Mtmr7
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal region of Human MTMR7 (aa 609-658).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54384, 9108
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.