Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MTMR8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109991
Description
Immunogen sequence: HVFSCQFGNF LGNCQKDRED LRVYEKTHSV WPFLVQRKPD FRNPLYKGFT MYGVLNPSTV PYNIQFWCGM YNRFDKGLQP KQSMLESLLE IKKQRAMLET DVHELEKKLK VRDEPPEEIC TCSQLGNILS Q.
This gene encodes a member of the myotubularin-related family and is part of the MTMR6 subgroup. Family members are dual-specificity phosphatases and the encoded protein contains a phosphoinositide-binding domain (PID) and a SET-interacting domain (SID). Defects in other family members have been found in myotubular myopathic diseases.
Specifications
| MTMR8 | |
| Polyclonal | |
| Unconjugated | |
| MTMR8 | |
| MTMR8; myotubularin related protein 8; myotubularin-related protein 8; Phosphatidylinositol-3,5-bisphosphate 3-phosphatase; Phosphatidylinositol-3-phosphate phosphatase | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 55613 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.58 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q96EF0 | |
| MTMR8 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 430-560 of human MTMR8 (NP_060147.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction