Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mtmr9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589003
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589003 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589003 Supplier Invitrogen™ Supplier No. PA589003
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEFAELIKTP RVDNVVLHRP FYPAVEGTLC LTGHHLILSS RQDNTEELWL LHSNIDAIDK RFVGSLGTII IKCKDFRIIQ LDIPGMEECL NIASSIEALS TLDSITLMYP FFYRPMFEVI EDGWHSFLPE QEFELYSSAT SEWRLSYVNK EFAVCPSYPP IVTVPKSIDD EALRKVATFR HGGRFPVLSY YHKKNGMVIM RSGQPLTGTN GRRCKEDEKL INATLRAGKR GYIIDTRSLN VAQQTRAKGG; Positive Samples: HepG2, HeLa, Jurkat, A-549, Mouse brain, Rat brain, Rat lung; Cellular Location: Cytoplasm.

This gene encodes a myotubularin-related protein that is atypical to most other members of the myotubularin-related protein family because it has no dual-specificity phosphatase domain. The encoded protein contains a double-helical motif similar to the SET interaction domain, which is thought to have a role in the control of cell proliferation. In mouse, a protein similar to the encoded protein binds with MTMR7, and together they dephosphorylate phosphatidylinositol 3-phosphate and inositol 1,3-bisphosphate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mtmr9
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Mtmr9
Gene Accession No. Q96QG7, Q9Z2D0
Gene Alias 9430075G12Rik; AA516943; AF073881; C8orf9; DKFZP434K171; Inactive phosphatidylinositol 3-phosphatase 9; LIP-STYX; MGC126672; mMTMH3; Mtmr3; MTMR8; MTMR9; myotubularin related protein 8; myotubularin related protein 9; myotubularin-related protein 9
Gene Symbols Mtmr9
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human MTMR9 (NP_0562732).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 210376, 282584, 66036
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.