Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MTMR9 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS17889R

Description
This gene encodes a myotubularin-related protein that is atypical to most other members of the myotubularin-related protein family because it has no dual-specificity phosphatase domain. The encoded protein contains a double-helical motif similar to the SET interaction domain, which is thought to have a role in the control of cell proliferation. In mouse, a protein similar to the encoded protein binds with MTMR7, and together they dephosphorylate phosphatidylinositol 3-phosphate and inositol 1,3-bisphosphate.Specifications
MTMR9 | |
Polyclonal | |
Unconjugated | |
Mtmr9 | |
9430075G12Rik; AA516943; AF073881; C8orf9; DKFZP434K171; Inactive phosphatidylinositol 3-phosphatase 9; LIP-STYX; MGC126672; mMTMH3; Mtmr3; MTMR8; MTMR9; myotubularin related protein 8; myotubularin related protein 9; Myotubularin-related protein 9 | |
Rabbit | |
Protein A | |
RUO | |
66036 | |
-20°C | |
Liquid |
Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q96QG7 | |
Mtmr9 | |
KLH conjugated synthetic peptide derived from human MTMR9. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction