Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MTNR1B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5102107
Description
Antibody detects endogenous levels of total MTR1B.
This gene encodes one of two high affinity forms of a receptor for melatonin, the primary hormone secreted by the pineal gland. This gene product is an integral membrane protein that is a G-protein coupled, 7-transmembrane receptor. It is found primarily in the retina and brain although this detection requires RT-PCR. It is thought to participate in light-dependent functions in the retina and may be involved in the neurobiological effects of melatonin.
Specifications
MTNR1B | |
Polyclonal | |
Unconjugated | |
MTNR1B | |
FGQTL2; LOW QUALITY PROTEIN: melatonin receptor type 1B; Mel1b; mel1b melatonin receptor; Mel-1b melatonin receptor; mel1b receptor; mel-1B-R; Mel1b-r; melatonin receptor 1B; melatonin receptor 1B variant b; melatonin receptor MEL1B; melatonin receptor type 1B; Mt2; MT2 melatonin receptor; MTNR1B; putative melatonin receptor 1B | |
Rabbit | |
Affinity chromatography | |
RUO | |
192646, 4544 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P49286, P49287 | |
MTNR1B | |
A synthesized peptide derived from human MTNR1B(Accession P49286), corresponding to amino acid residues S6-V56. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction