Learn More
Invitrogen™ MTNR1B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA532935
Description
Percent identity with other species by BLAST analysis: Human (100%); Gorilla (94%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
This gene encodes one of two high affinity forms of a receptor for melatonin, the primary hormone secreted by the pineal gland. This gene product is an integral membrane protein that is a G-protein coupled, 7-transmembrane receptor. It is found primarily in the retina and brain although this detection requires RT-PCR. It is thought to participate in light-dependent functions in the retina and may be involved in the neurobiological effects of melatonin.
Specifications
| MTNR1B | |
| Polyclonal | |
| Unconjugated | |
| MTNR1B | |
| FGQTL2; LOW QUALITY PROTEIN: melatonin receptor type 1B; Mel1b; mel1b melatonin receptor; Mel-1b melatonin receptor; mel1b receptor; mel-1B-R; Mel1b-r; melatonin receptor 1B; melatonin receptor 1B variant b; melatonin receptor MEL1B; melatonin receptor type 1B; Mt2; MT2 melatonin receptor; MTNR1B; putative melatonin receptor 1B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4544 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P49286 | |
| MTNR1B | |
| Synthetic 16 amino acid peptide from N-terminal extracellular domain of human MTNR1B. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.