Learn More
Invitrogen™ MTNR1B Polyclonal Antibody

Description
This target displays homology in the following species: Human: 100%; Pig: 79%; Rabbit: 79% The immunogen sequence of amino acid 6-55 is: SFANCCEAGG WAVRPGWSGA GSARPSRTPR PPWVAPALSA VLIVTTAVDV.
Specifications
Specifications
| Antigen | MTNR1B |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | MTNR1B |
| Gene Accession No. | P49286 |
| Gene Alias | FGQTL2; LOW QUALITY PROTEIN: melatonin receptor type 1B; Mel1b; mel1b melatonin receptor; Mel-1b melatonin receptor; mel1b receptor; mel-1B-R; Mel1b-r; melatonin receptor 1B; melatonin receptor 1B variant b; melatonin receptor MEL1B; melatonin receptor type 1B; Mt2; MT2 melatonin receptor; MTNR1B; putative melatonin receptor 1B |
| Gene Symbols | MTNR1B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.