Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTNR1B Polyclonal Antibody, HRP, Bioss
SDP

Catalog No. 502279598
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-9598 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-9598 Supplier Bioss Supplier No. BS21511RHRP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MTNR1B Polyclonal antibody specifically detects MTNR1B in Human, Mouse samples. It is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen MTNR1B
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate HRP
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene MTNR1B
Gene Accession No. P49286, Q8CIQ6
Gene Alias FGQTL2; LOW QUALITY PROTEIN: melatonin receptor type 1B; Mel1b; mel1b melatonin receptor; Mel-1b melatonin receptor; mel1b receptor; mel-1B-R; Mel1b-r; melatonin receptor 1B; melatonin receptor 1B variant b; melatonin receptor MEL1B; melatonin receptor type 1B; Mt2; MT2 melatonin receptor; MTNR1B; putative melatonin receptor 1B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MTNR1B (271-364).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 244701, 4544
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.