Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTNR1B, Rabbit anti-Human, Polyclonal, Millipore Sigma™

Catalog No. ABT189025UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABT189025UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABT189025UG Supplier MilliporeSigma Supplier No. ABT189025UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-MTNR1B Antibody, Cat. No. ABT1890, is a rabbit polyclonal antibody that detects MTNR1B and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.

Melatonin receptor type 1B (UniProt: P49286; also known as Mel-1B-R, Mel1b receptor) is encoded by the MTNR1B gene (Gene ID: 4544) in human. Mel-1B-R is a member of the G-protein coupled 1 family that is predominantly expressed in retina and to lesser extent in brain tissue. . Mel-1B-R activity is mediated by pertussis toxin sensitive G proteins that inhibit adenylate cyclase activity. Mel-1B-R mediates the reproductive and circadian actions of melatonin. Melatonin is shown to be involved in body mass regulation in mammals and this effect is suggest to be mediated by a direct action of melatonin on Mel-1B receptors on brown adipocytes. Melatonin is reported to act as a chemopreventive agent and its levels inversely correlate with the risk of developing cancer. In this regard melatonin induces p38-dependent phosphorylation of both p53 and histone H2AX. Activation of the p53-dependent DNA damage response by melatonin is mediated by both Mel-1A and Mel-1B receptors. The absence of either receptor impairs melatonin's ability to reduce both cell proliferation and clonogenic potential of cancer cells. A variant in MTNR1B gene is reported to worsen the deleterious effect of melatonin on glucose tolerance in humans. (Ref.: Brydon, L. et al. (2001). Endocrinology. 142(10):4264-4271; Santoro, R., et al. (2013). Carcinogenesis. 34(5):1051-1061).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTNR1B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected MTNR1B in human retina tissue.
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Melatonin receptor type 1B;Mel-1B-R
Gene Symbols MTNR1B
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 20 amino acids from the N-terminal region of human Melatonin receptor type 1B.
Molecular Weight of Antigen 40.19 kDa calculated.
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects Melatonin receptor type 1B in human.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.