Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556408
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556408 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556408 Supplier Invitrogen™ Supplier No. PA556408
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RPDNADSRLT LRGYKDAGCV SQQRYERACW MKSSLEEGIS VLKSIEFLSS KWKKLIPEAS ISTSRSLPVR ALDVLKYE Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 79%.

MTO1 encodes a mitochondrial protein thought to be involved in mitochondrial tRNA modification. The encoded protein may also play a role in the expression of the non-syndromic and aminoglycoside-induced deafness phenotypes associated with a specific mutation in the mitochondrial 12S rRNA gene. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTO1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MTO1
Gene Accession No. Q9Y2Z2
Gene Alias 2310039H01Rik; 5730419A02Rik; CGI02; CGI-02; COXPD10; homolog of yeast Mto1; LOW QUALITY PROTEIN: protein MTO1 homolog, mitochondrial; mitochondrial MTO1-3; mitochondrial translation optimization 1 homolog; mitochondrial translation optimization 1 homolog (S. cerevisiae); mitochondrial tRNA translation optimization 1; MTO1; Protein MTO1 homolog, mitochondrial
Gene Symbols MTO1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MTO1. Recombinant protein control fragment (Product #RP-94831).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25821
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.