Learn More
mTOR Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300503AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and Rat
Mammalian target of rapamycin (mTOR) is a serine/threonine kinase that plays a key role in cell growth, cell proliferation, and protein synthesis. mTOR mediates phosphoinositide 3-kinase and Akt/PKB signaling, resulting in phosphorylation of 4EBP1, and initiation of mRNA translation. A second pathway involves regulation of ribosomal S6 kinase, which affects ribosome biogenesis and translation elongation. Mutations affecting the gene results in Smith-Kingsmore syndrome.Specifications
mTOR | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P42345 | |
MTOR | |
Between 200 and 250 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Western Blot | |
Unconjugated | |
MTOR | |
MTOR, m-TOR, Mechanistic Target Of Rapamycin, Mammalian Target Of Rapamycin, FRAP, FRAP1, FRAP2, FK506 binding protein 12-rapamycin associated protein, FKBP-Rapamycin Associated Protein, Rapamycin Target Protein, Serine/Threonine-Protein Kinase MTOR, EC 2.7.11.1, RAFT1, RAPT1, mTORC1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2475 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.