Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mTOR Polyclonal Antibody, QED Bioscience™
SDP

Catalog No. 502277490
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7490 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-7490 Supplier Leinco Technologies Inc Supplier No. 13036100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

mTOR Polyclonal antibody specifically detects mTOR in Human, Mouse, Rat samples. It is validated for Immunoprecipitation, Western Blot

Specifications

Antigen mTOR
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene MTOR
Gene Accession No. P42345, P42346, Q9JLN9
Gene Alias 2610315D21Rik; AI327068; angiopoietin-like factor CDT6; FK506 binding protein 12-rapamycin associated protein 1; FK506 binding protein 12-rapamycin associated protein 2; FK506-binding protein 12-rapamycin complex-associated protein 1; FKBP12-rapamycin complex-associated protein; FKBP12-rapamycin complex-associated protein 1; FKBP-rapamycin associated protein; FKBP-rapamycin associated protein (FRAP); FKBP-rapamycin-associated protein FRAP; flat; FRAP; FRAP1; FRAP2; Mammalian target of rapamycin; mechanistic target of rapamycin; mechanistic target of rapamycin (serine/threonine kinase); mechanistic target of rapamycin kinase; Mtor; m-TOR; mTORC1; Raft1; Rapamycin and FKBP12 target 1; rapamycin and FKBP12 target-1 protein; rapamycin associated protein FRAP2; rapamycin target protein 1; RAPT1; serine/threonine-protein kinase mTOR; SKS
Host Species Rabbit
Immunogen Synthetic peptide representing a portion of the protein encoded in part by exons 5 and 6 (LocusLink ID 1981).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2475, 56717, 56718
Target Species Human, Mouse, Rat
Content And Storage 2-8°C
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.