Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mTOR Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501559362
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-9362 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-9362 Supplier Bethyl Laboratories, Inc Supplier No. A301143A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat

Mammalian target of rapamycin (mTOR) is a serine/threonine kinase that plays a key role in cell growth, cell proliferation, and protein synthesis. mTOR mediates phosphoinositide 3-kinase and Akt/PKB signaling, resulting in phosphorylation of 4EBP1, and initiation of mRNA translation. A second pathway involves regulation of ribosomal S6 kinase, which affects ribosome biogenesis and translation elongation. Mutations affecting the gene results in Smith-Kingsmore syndrome.

Specifications

Antigen mTOR
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene MTOR
Gene Accession No. P42345, Q9JLN9
Gene Alias MTOR, m-TOR, Mechanistic Target Of Rapamycin, Mammalian Target Of Rapamycin, FRAP, FRAP1, FRAP2, FK506 binding protein 12-rapamycin associated protein, FKBP-Rapamycin Associated Protein, Rapamycin Target Protein, Serine/Threonine-Protein Kinase MTOR, EC 2.7.11.1, RAFT1, RAPT1, mTORC1
Gene Symbols Mtor
Host Species Rabbit
Immunogen Between 1350 and 1400
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2475, 56717
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.