Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115599
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115599 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115599 Supplier Invitrogen™ Supplier No. PA5115599
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MTR.

MTR encodes the enzyme 5-methyltetrahydrofolate-homocysteine methyltransferase. This enzyme, also known as cobalamin-dependent methionine synthase, catalyzes the final step in methionine biosynthesis. Mutations in MTR have been identified as the underlying cause of methylcobalamin deficiency complementation group G.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTR
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene MTR
Gene Accession No. A6H5Y3, Q99707, Q9Z2Q4
Gene Alias 5-methyltetrahydrofolate-homocysteine methyltransferase; 5-methyltetrahydrofolate--homocysteine methyltransferase; 5-methyltetrahydrofolate-homocysteine methyltransferase 1; AI894170; cblG; cobalamin-dependent methionine synthase; D830038K18Rik; FLJ33168; FLJ43216; FLJ45386; HMAG; methionine synthase; Methionine synthase-like protein; methioninesynthase; MS; MTR; Vitamin-B12 dependent methionine synthase
Gene Symbols MTR
Host Species Rabbit
Immunogen A synthesized peptide derived from human MTR(Accession Q99707), corresponding to amino acid residues M267-V317.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 238505, 4548, 81522
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.