Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTRES1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144192
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144192 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144192 Supplier Invitrogen™ Supplier No. PA5144192
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MTRES1 Polyclonal Antibody for IHC (P), ELISA

MTRES1 is a mitochondrial RNA-binding protein involved in mitochondrial transcription regulation. It functions as a protective factor to maintain proper mitochondrial RNA level during stress. It acts at the transcription level and its protective function depends on its RNA binding ability.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTRES1
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene MTRES1
Gene Accession No. Q9P0P8
Gene Alias C6orf203; chromosome 6 open reading frame 203; HSPC230; Mitochondrial transcription rescue factor 1; MTRES1; PRED31; uncharacterized protein C6orf203
Gene Symbols C6orf203, MTRES1
Host Species Rabbit
Immunogen Recombinant Human Uncharacterized protein C6orf203 protein (11-240AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51250
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.