Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTRF1 Monoclonal Antibody (1D5)
GREENER_CHOICE

Catalog No. PIMA524442
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524442 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524442 Supplier Invitrogen™ Supplier No. MA524442
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MNRHLCVWLF RHPSLNGYLQ CHIQLHSHQF RQIHLDTRLQ VFRQNRNCIL HLLSKNWSRR YCHQDTKMLW KHKALQKYME NLSKEYQTLE QCLQHIPVNE.

Mitochondrial peptide chain release factor that directs the termination of translation in response to the peptide chain non-cognate termination stop codons AGG and AGA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTRF1
Applications ELISA
Classification Monoclonal
Clone 1D5
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene MTRF1
Gene Accession No. O75570
Gene Alias A830062K05Rik; mitochondrial translational release factor 1; mitochontrial peptide chain release factor 1; MRF1; MRF-1; Mtrf1; MtRF-1; MTTRF1; Peptide chain release factor 1, mitochondrial; RF1
Gene Symbols MTRF1
Host Species Mouse
Immunogen MTRF1 (NP_004285.2, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9617
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.