Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTRF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560319
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560319 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560319 Supplier Invitrogen™ Supplier No. PA560319
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IAFRVLRARL YQQIIEKDKR QQQSARKLQV GTRAQSERIR TYNFTQDRVS DHRIAYEVRD IKEFLCGGKG LDQLIQRLLQ SADEEAIAEL LDEHL Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 89%.

Mitochondrial peptide chain release factor that directs the termination of translation in response to the peptide chain non-cognate termination stop codons AGG and AGA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTRF1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MTRF1
Gene Accession No. O75570
Gene Alias A830062K05Rik; mitochondrial translational release factor 1; mitochontrial peptide chain release factor 1; MRF1; MRF-1; Mtrf1; MtRF-1; MTTRF1; Peptide chain release factor 1, mitochondrial; RF1
Gene Symbols MTRF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MTRF1. Recombinant protein control fragment (Product #RP-97929).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9617
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.