Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MTRF1L Antibody (OTI1D11), PE/Cy5.5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272814PECY55
Description
MTRF1L Monoclonal specifically detects MTRF1L in Human samples. It is validated for Flow Cytometry.Specifications
MTRF1L | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
MTRF1L | |
Full length recombinant protein of human MTRF1L (NP_061914) produced in HEK293T cells. | |
0.1 mL | |
54516.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
OTI1D11 | |
Flow Cytometry | |
mitochondrial translational release factor 1-likemitochondrial, MRF1L, MTRF1A | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction